1-Butyl-3-methylimidazoliumtetrafluoroborat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Butyl-1-methyl-1H-imidazol-3-ium tetrafluoroborate, 1-n-Butyl-3-methylimidazolium tetrafluoroborate, [BMIM] [BF4]
L19087.09
L19087.18
L19087.09EA
132
EUR
L19087.09
L19087.18
1-Butyl-3-methylimidazoliumtetrafluoroborat ≥98%
1-Butyl-3-methylimidazoliumtetrafluoroborate
Purity: 98+%
CAS No.: 174501-65-6
Molecular Formula: C8H15BF4N2
Formula Weight: 226.03
Flash Point: None
Refractive Index: 1.4220
MDL No.: MFCD03095449
Formel:
C₈H₁₅BF₄N₂ Molecular Weight: 226.02 g/mol Siedepunkt: ∼200 °C (1013 hPa) Schmelzpunkt: –71 °C Dichte: 1.210 Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095449 CAS-Nummer: 174501-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...