Dichloro-(p-cymol)-ruthenium(II) Dimer 98%

Lieferant: Thermo Fisher Scientific
Gefahr

L19126.04 L19126.09 L19126.ME
L19126.04EA 110 EUR
L19126.04 L19126.09 L19126.ME
Dichloro-(p-cymol)-ruthenium(II) Dimer 98%
Dichloro(p-cymene)ruthenium(II) dimer

Purity: 98% CAS No.: 52462-29-0 Molecular Formula: C20H28Cl4Ru2 Formula Weight: 612.40 MDL No.: MFCD00064793

Formel: C₂₀H₂₈Cl₄Ru₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00064793
CAS-Nummer: 52462-29-0
EINECS: 435-530-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR