2-Chlor-3,6-difluorbenzylamin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L19140.03
L19140.03EA
74.9
EUR
L19140.03
2-Chlor-3,6-difluorbenzylamin 97%
2-Chlor-3,6-difluorbenzylamin
Purity: 97%
CAS No.: 261762-45-2
Molecular Formula: C7H6ClF2N
Formula Weight: 177.58
Refractive Index: 1.5200
Formel:
C₇H₆ClF₂N Molecular Weight: 177.58 g/mol Siedepunkt: 207 Schmelzpunkt: 73 Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631449 CAS-Nummer: 261762-45-2 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...