2-(Difluormethoxy)benzylamin 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (2-(Difluoromethoxy)phenyl)methanamine

L19165.03
L19165.03EA 80.8 EUR
L19165.03
2-(Difluormethoxy)benzylamin 97%
2-(Difluormethoxy)benzylamin

Purity: 97% CAS No.: 243863-36-7 Molecular Formula: C8H9F2NO Formula Weight: 173.16 Refractive Index: 1.4910

Formel: C₈H₉F₂NO
Molecular Weight: 173,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236229
CAS-Nummer: 243863-36-7
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR