3-Chlor-2-fluorbenzylamin 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (3-Chloro-2-fluorophenyl)methanamine

L19227.03
L19227.03EA 104 EUR
L19227.03
3-Chlor-2-fluorbenzylamin 97%
3-Chlor-2-fluorbenzylamin

Purity: 97% CAS No.: 72235-55-3 Molecular Formula: C7H7ClFN Formula Weight: 159.59 Refractive Index: 1.5405

Formel: C₇H₇ClFN
Molecular Weight: 159,59 g/mol
Siedepunkt: 211 °C (1013 hPa)
Schmelzpunkt: 60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631391
CAS-Nummer: 72235-55-3
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR