3-(Benzyloxy)phenylisothiocyanat ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Benzyloxyphenylisothiocyanate

L19242.03 L19242.MD
L19242.03EA 124 EUR
L19242.03 L19242.MD
3-(Benzyloxy)phenylisothiocyanat ≥97%
3-(Benzyloxy)phenylisothiocyanat

Purity: 97% CAS No.: 206559-36-6 Molecular Formula: C14H11NOS Formula Weight: 241.31 Melting Point: 49-51° UN No.: 2923 MDL No.: MFCD00060374

Formel: C₁₄H₁₁NOS
Schmelzpunkt: 49…51 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060374
CAS-Nummer: 206559-36-6
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR