2-Brom-5-iodbenzoesäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L19247.06
L19247.06EA
121
EUR
L19247.06
2-Brom-5-iodbenzoesäure ≥97%
2-Brom-5-iodbenzoesäure
Purity: 97%
CAS No.: 25252-00-0
Molecular Formula: C7H4BrIO2
Formula Weight: 326.92
Melting Point: 174-176°
MDL No.: MFCD00079716
Formel:
C₇H₄BrIO₂ Schmelzpunkt: 174…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079716 CAS-Nummer: 25252-00-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...