4-Iodbenzylalkohol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (4-Iodophenyl)methanol

L19278.03 L19278.06
L19278.03EA 40.2 EUR
L19278.03 L19278.06
4-Iodbenzylalkohol ≥97%
4-Iodbenzylalkohol

Purity: 97% CAS No.: 18282-51-4 Molecular Formula: C7H7IO Formula Weight: 234.04 Melting Point: 72-74° Beilstein Reference: 1931621

Formel: C₇H₇IO
Molecular Weight: 234.04 g/mol
Schmelzpunkt: 72…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01732720
CAS-Nummer: 18282-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR