3-Chlor-4-fluor-5-nitrobenzotrifluorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-Chloro-2-fluoro-3-nitro-5-(trifluoromethyl)benzene, 3-Chloro-4-fluoro-5-nitrobenzotrifluoride
L19322.06
L19322.06EA
97.4
EUR
L19322.06
3-Chlor-4-fluor-5-nitrobenzotrifluorid ≥98%
3-Chlor-4-fluor-5-nitrobenzotrifluorid
Purity: 98%
CAS No.: 101646-02-0
Molecular Formula: C7H2ClF4NO2
Formula Weight: 243.55
Flash Point:
Formel:
C₇H₂ClF₄NO₂ Molecular Weight: 243.54 g/mol Siedepunkt: 200 °C Dichte: 1.61 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00674108 CAS-Nummer: 101646-02-0 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...