N-Boc-L-isoleucinol 95%
Lieferant: Thermo Fisher Scientific
L19351.06
L19351.06EA
180
EUR
L19351.06
N-Boc-L-isoleucinol 95%
N-Boc-L-isoleucinol
Purity: 95%
CAS No.: 106946-74-1
Molecular Formula: C11H23NO3
Formula Weight: 217.31
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4530
Boiling Point: 121-126°/0.6mm
Beilstein Reference: 3604099
MDL No.: MFCD00235929
Formel:
C₁₁H₂₃NO₃ Siedepunkt: 121…126 °C (0,6 mmHg) |
MDL:
MFCD00235929 CAS-Nummer: 106946-74-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...