N-Boc-L-leucinol 97%

Lieferant: Thermo Fisher Scientific

Synonyme: (S)-tert-Butyl (1-hydroxy-4-methylpentan-2-yl)carbamate, Boc-Leucinol

L19352.03 L19352.06
L19352.03EA 62.4 EUR
L19352.03 L19352.06
N-Boc-L-leucinol 97%
N-Boc-L-leucinol

Purity: 97% CAS No.: 82010-31-9 Molecular Formula: C11H23NO3 Formula Weight: 217.31 Flash Point: 110°(230°F) Refractive Index: 1.4500 Boiling Point: 212-214° Beilstein Reference: 4178460 Optical Rotation: -27.5° (c=2 in methanol) MDL No.: MFCD00076950

Formel: C₁₁H₂₃NO₃
Siedepunkt: 212…214 °C
Dichte: 0,980
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00076950
CAS-Nummer: 82010-31-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR