1-Allylpiperazin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L19391.06
L19391.14
L19391.06EA
79.4
EUR
L19391.06
L19391.14
1-Allylpiperazin ≥98%
1-Allylpiperazin
Purity: 98+%
CAS No.: 13961-36-9
Molecular Formula: C7H14N2
Formula Weight: 126.20
Flash Point: 51°(123°F)
Refractive Index: 1.4830
Boiling Point: 180-182°
Beilstein Reference: 507447
UN No.: 2920
MDL No.: MFCD00167970
Formel:
C₇H₁₄N₂ Siedepunkt: 180…182 °C Dichte: 0,902 Flammpunkt: 51 °C (123 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00167970 CAS-Nummer: 13961-36-9 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...