5-Bromo-2-methyl-2-pentene 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L19406.03
L19406.06
L19406.03EA
68.7
EUR
L19406.03
L19406.06
5-Bromo-2-methyl-2-pentene 97%
5-Bromo-2-methyl-2-pentene
Purity: 97%
CAS No.: 2270-59-9
Molecular Formula: C6H11Br
Formula Weight: 163.06
Storage Temperature: 2°C to 8°C
Flash Point: 23°(73°F)
Refractive Index: 1.4775
Boiling Point: 50°/20mm
Beilstein Reference: 1735749
UN No.: UN1993
MDL No.: MFCD00009887
Formel:
C₆H₁₁Br Siedepunkt: 50 °C (20 mmHg) Dichte: 1,219 Flammpunkt: 23 °C (73 °F) |
MDL:
MFCD00009887 CAS-Nummer: 2270-59-9 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...