1,3-Benzoldithiol ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 626-04-0
Molecular Formula: C6H6S2
Formula Weight: 142.24
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₆H₆S₂ Siedepunkt: 244 °C Schmelzpunkt: 27 °C Dichte: 1,24 Flammpunkt: >112 °C (234 °F) |
MDL:
MFCD00004842 CAS-Nummer: 626-04-0 EINECS: 210-925-3 UN: 3335 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...