4-(Trifluormethyl)piperidin Hydrochlorid ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L19441.MD
L19441.MDEA
46.6
EUR
L19441.MD
4-(Trifluormethyl)piperidin Hydrochlorid ≥97%
4-(Trifluormethyl)piperidin Hydrochlorid
Purity: 97%
CAS No.: 155849-49-3
Molecular Formula: C6H10F3N.HCl
Formula Weight: 189.61
Melting Point: ≈160° dec.
Beilstein Reference: 1341222
Formel:
C₆H₁₁ClF₃N Molecular Weight: 189.61 g/mol Schmelzpunkt: ∼160 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04971993 CAS-Nummer: 155849-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...