6-Chlorpicolinsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Chloropyridine-2-carboxylic acid, 2-Chloropyridine-6-carboxylic acid
L19448.06
L19448.06EA
109
EUR
L19448.06
6-Chlorpicolinsäure ≥98%
6-Chlorpicolinsäure
Purity: 98%
CAS No.: 4684-94-0
Molecular Formula: C6H4ClNO2
Formula Weight: 157.56
Melting Point: 194°
Beilstein Reference: 115849
Formel:
C₆H₄ClNO₂ Molecular Weight: 157,56 g/mol Schmelzpunkt: 194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00155390 CAS-Nummer: 4684-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...