N-Boc-1,3-propanediamine hydrochloride 98%
Lieferant: Thermo Fisher Scientific
L19486.03
L19486.03EA
92.2
EUR
L19486.03
N-Boc-1,3-propanediamine hydrochloride 98%
N-Boc-1,3-propanediamine hydrochloride
Purity: 98%
CAS No.: 127346-48-9
Molecular Formula: (CH3)3COCONHCH2CH2CH2NH2.HCl
Formula Weight: 210.71
Melting Point: 159-161°
Beilstein Reference: 3627894
MDL No.: MFCD01076598
Formel:
C₈H₁₉ClN₂O₂ Schmelzpunkt: 159…161 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01076598 CAS-Nummer: 127346-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...