[4-(Methylsulfonyl)phenyl]essigsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

L19504.06 L19504.14
L19504.06EA 43 EUR
L19504.06 L19504.14
[4-(Methylsulfonyl)phenyl]essigsäure 97%
[4-(Methylsulfonyl)phenyl]essigsäure

Purity: 97% CAS No.: 90536-66-6 Molecular Formula: C9H10O4S Formula Weight: 214.24 Melting Point: 134-138° Beilstein Reference: 2646649 MDL No.: MFCD00216495

Formel: C₉H₁₀O₄S
Schmelzpunkt: 134…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216495
CAS-Nummer: 90536-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR