4-Fluor-2-methylbenzolsulfonylchlorid ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Fluor-2-methylbenzol-1-sulfonylchlorid, 4-Fluor-2-methylbenzolsulfochlorid, 4-Fluor-2-methylbenzolsulfonsäurechlorid

L19517.03 L19517.06
L19517.03EA 36.9 EUR
L19517.03 L19517.06
4-Fluor-2-methylbenzolsulfonylchlorid ≥97%
4-Fluor-2-methylbenzolsulfonylchlorid

Purity: 97% CAS No.: 7079-48-3 Molecular Formula: C7H6ClFO2S Formula Weight: 208.64 Flash Point: 110°(230°F) Refractive Index: 1.5370 Boiling Point: 245-246° Beilstein Reference: 6720115 UN No.: 3265 MDL No.: MFCD03094390

Formel: C₇H₆ClFO₂S
Molecular Weight: 208.64 g/mol
Siedepunkt: 245…246 °C
Dichte: 1.433
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD03094390
CAS-Nummer: 7079-48-3
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR