2-Brom-3-methylpyridin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Bromo-3-picoline

L19522.06
L19522.06EA 62.8 EUR
L19522.06
2-Brom-3-methylpyridin ≥98%
2-Brom-3-methylpyridin

Purity: 98+% CAS No.: 3430-17-9 Molecular Formula: C6H6BrN Formula Weight: 172.03 Flash Point: 110°(230°F) Refractive Index: 1.5695 Boiling Point: 218-219° Beilstein Reference: 107896 MDL No.: MFCD00239380

Formel: C₆H₆BrN
Molecular Weight: 172,02 g/mol
Siedepunkt: 218…219 °C
Dichte: 1,544
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00239380
CAS-Nummer: 3430-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR