3,3-Diethoxy-1-propylboronsäurepinakolester ≥97%

Lieferant: Thermo Scientific

L19532.03
L19532.03EA 209 EUR
L19532.03
3,3-Diethoxy-1-propylboronsäurepinakolester ≥97%
3,3-Diethoxy-1-propylboronsäurepinakolester

Purity: 97% CAS No.: 165904-27-8 Molecular Formula: C13H27BO4 Formula Weight: 258.17 Refractive Index: 1.4280

Formel: C₁₃H₂₇BO₄
Molecular Weight: 258,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03788723
CAS-Nummer: 165904-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR