2-Amino-4-(trifluormethyl)pyridin 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-(Trifluoromethyl)pyridin-2-amine, 4-(Trifluoromethyl)-2-pyridylamine

L19548.ME
L19548.MEEA 185 EUR
L19548.ME
2-Amino-4-(trifluormethyl)pyridin 99%
2-Amino-4-(trifluoromethyl)pyridine

Purity: 99% CAS No.: 106447-97-6 Molecular Formula: C6H5F3N2 Formula Weight: 162.11 Melting Point: 71-74° Beilstein Reference: 7869677 UN No.: 2811 MDL No.: MFCD00077455

Formel: C₆H₅F₃N₂
Molecular Weight: 162,11 g/mol
Schmelzpunkt: 71…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077455
CAS-Nummer: 106447-97-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR