4,4-Difluorpiperidin Hydrochlorid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4,4-Difluoropiperidine HCl

L19555.03
L19555.03EA 202 EUR
L19555.03
4,4-Difluorpiperidin Hydrochlorid ≥98%
4,4-Difluorpiperidin Hydrochlorid

Purity: 98% CAS No.: 144230-52-4 Molecular Formula: C5H9F2N.HCl Formula Weight: 157.59 Melting Point: ca 172° Beilstein Reference: 6812163

Formel: C₅H₁₀ClF₂N
Molecular Weight: 157,59 g/mol
Schmelzpunkt: ∼ 172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095381
CAS-Nummer: 144230-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR