4-Chlor-3-picolin Hydrochlorid ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L19558.03
L19558.03EA
111
EUR
L19558.03
4-Chlor-3-picolin Hydrochlorid ≥97%
4-Chlor-3-picolin Hydrochlorid
Purity: 97%
CAS No.: 19524-08-4
Molecular Formula: C6H6ClN.HCl
Formula Weight: 164.04
Odour: Odourless
Melting Point: ca 185°
Beilstein Reference: 107953
MDL No.: MFCD03092890
Formel:
C₆H₇Cl₂N Schmelzpunkt: ∼ 185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03092890 CAS-Nummer: 19524-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...