Amberlite® XAD-16 Ionenaustauscher
Lieferant: Thermo Fisher Scientific
L19565.22
L19565.36
L19565.22EA
53.9
EUR
L19565.22
L19565.36
Amberlite® XAD-16 Ionenaustauscher
Amberlite® XAD-16
CAS No.: 104219-63-8
MDL No.: MFCD00145831
Formel:
C₁₀H₁₀ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00145831 CAS-Nummer: 104219-63-8 Merck Index: 14,382 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...