3-Amino-2-nitropyridin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Nitropyridin-3-amin, 2-Nitro-3-pyridylamin, 2-Nitropyridin-3-ylamin

L19619.06
L19619.06EA 96 EUR
L19619.06
3-Amino-2-nitropyridin 97%
3-Amino-2-nitropyridin

Purity: 97% CAS No.: 13269-19-7 Molecular Formula: C5H5N3O2 Formula Weight: 139.11 Melting Point: 191-193° Beilstein Reference: 124467

Formel: C₅H₅N₃O₂
Molecular Weight: 139,11 g/mol
Schmelzpunkt: 191…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00044102
CAS-Nummer: 13269-19-7
EINECS: 236-260-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR