5-Isochinolincarbonsäure 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: Isoquinoline-5-carboxylic acid

L19637.MD
L19637.MDEA 96 EUR
L19637.MD
5-Isochinolincarbonsäure 96%
5-Isoquinolinecarboxylic acid

Purity: 96% CAS No.: 27810-64-6 Molecular Formula: C10H7NO2 Formula Weight: 173.17 Melting Point: 278-281° Boiling Point: 60-62°/11mm MDL No.: MFCD03788744

Formel: C₁₀H₇NO₂
Molecular Weight: 173,17 g/mol
Siedepunkt: 60…62 °C (11 mmHg)
Schmelzpunkt: 278…281 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788744
CAS-Nummer: 27810-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR