8-Chinolinboronsäure ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

L19640.03 L19640.MD
L19640.03EA 126 EUR
L19640.03 L19640.MD
8-Chinolinboronsäure ≥99%
8-Quinolineboronic acid

Purity: 99% CAS No.: 86-58-8 Molecular Formula: C9H8BNO2 Formula Weight: 172.98 Melting Point: 300° Beilstein Reference: 147522 Merck Reference: 14,8069 MDL No.: MFCD01114698

Formel: C₉H₈BNO₂
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01114698
CAS-Nummer: 86-58-8
Merck Index: 14,8069

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR