(E)-α,β-Styroldiboronsäurebis(pinakol)ester ≥98%
Lieferant: Thermo Scientific
L19651.03
L19651.03EA
80.3
EUR
L19651.03
(E)-α,β-Styroldiboronsäurebis(pinakol)ester ≥98%
(E)-α,β-Styroldiboronsäurebis(pinakol)ester
Purity: 98%
CAS No.: 173603-23-1
Molecular Formula: C20H30B2O4
Formula Weight: 356.08
Melting Point: 69-73°
Beilstein Reference: 8855562
Formel:
C₂₀H₃₀B₂O₄ Molecular Weight: 356,08 g/mol Schmelzpunkt: 69…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683500 CAS-Nummer: 173603-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...