tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate 97%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Boc-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-(Boc-amino)phenylboronic acid Pinacol ester, tert-Butyl [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate, 4-[(tert-Butoxycarbonyl)amino]phenylboronic acid Pinacol ester, 4-Boc-aminophenylboronic acid, pinacol ester, N-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, tert-Butyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate, [4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamic acid tert-butyl ester, 4-(Boc-amino)benzeneboronic acid pinacol ester

L19653.03 L19653.06
L19653.03EA 75 EUR
L19653.03 L19653.06
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate 97%
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate

Purity: 97% CAS No.: 330793-01-6 Molecular Formula: C17H26BNO4 Formula Weight: 319.21 Melting Point: 169-170° MDL No.: MFCD02179439

Formel: C₁₇H₂₆BNO₄
Schmelzpunkt: 169…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179439
CAS-Nummer: 330793-01-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR