1H-Pyrazol-4-boronsäurepinakolester ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L19654.03 L19654.06
L19654.03EA 74.8 EUR
L19654.03 L19654.06
1H-Pyrazol-4-boronsäurepinakolester ≥98%
1H-Pyrazol-4-boronsäurepinakolester

Purity: 98% CAS No.: 269410-08-4 Molecular Formula: C9H15BN2O2 Formula Weight: 194.04 Melting Point: 145-149° MDL No.: MFCD03453063

Formel: C₉H₁₅BN₂O₂
Schmelzpunkt: 145…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03453063
CAS-Nummer: 269410-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR