2-Fluor-3-methoxyphenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Fluoro-3-methoxybenzeneboronic acid, (2-Fluoro-3-methoxyphenyl)boronic acid

L19655.06
L19655.06EA 75.6 EUR
L19655.06
2-Fluor-3-methoxyphenylboronsäure ≥97%
2-Fluor-3-methoxyphenylboronsäure

Purity: 97% CAS No.: 352303-67-4 Molecular Formula: C7H8BFO3 Formula Weight: 169.95 Beilstein Reference: 9191510 MDL No.: MFCD02094728

Formel: C₇H₈BFO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02094728
CAS-Nummer: 352303-67-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR