4-Fluor-3-methoxybenzoesäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Methoxy-4-fluorobenzoic acid

L19673.06
L19673.06EA 151 EUR
L19673.06
4-Fluor-3-methoxybenzoesäure 95%
4-Fluoro-3-methoxybenzoic acid

Purity: 95% CAS No.: 82846-18-2 Molecular Formula: C8H7FO3 Formula Weight: 170.14 Melting Point: 206-208° Beilstein Reference: 4178071

Formel: C₈H₇FO₃
Molecular Weight: 170,14 g/mol
Schmelzpunkt: 206…208 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00272134
CAS-Nummer: 82846-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR