(1-Hydroxyethyl)ferrocen ≥99%

Lieferant: Thermo Fisher Scientific

L19681.03 L19681.06
L19681.03EA 17.3 EUR
L19681.03 L19681.06
(1-Hydroxyethyl)ferrocen ≥99%
1-Hydroxyethylferrocene

Purity: 99% CAS No.: 1277-49-2 Molecular Formula: C12H14FeO Formula Weight: 230.09 Flash Point: 100°(212°F) Melting Point: 77-79° MDL No.: MFCD00013752

Formel: C₁₂H₁₄FeO
Schmelzpunkt: 77…79 °C
Flammpunkt: >100 °C (212 °F)
MDL: MFCD00013752
CAS-Nummer: 1277-49-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR