(3R)-1-Methyl-3-pyrrolidinol ≥99%, ee 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: (R)-3-Hydroxy-1-methylpyrrolidine, (R)-(-)-1-Methyl-3-pyrrolidinol, (R)-3-Hydroxy-1-methyl-pyrrolidine

L19694.03
L19694.03EA 74.6 EUR
L19694.03
(3R)-1-Methyl-3-pyrrolidinol ≥99%, ee 99%
(R)-1-Methyl-3-pyrrolidinol

Purity: 99% CAS No.: 104641-60-3 Molecular Formula: C5H11NO Formula Weight: 101.15 Flash Point: 70°(158°F) Refractive Index: 1.4640 Boiling Point: 103-106°/35mm Beilstein Reference: 4349393 MDL No.: MFCD03788747

Formel: C₅H₁₁NO
Molecular Weight: 101,15 g/mol
Siedepunkt: 103…106 °C (35 mmHg)
Dichte: 0,920
Flammpunkt: 70 °C (158 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD03788747
CAS-Nummer: 104641-60-3
EINECS: 000-000-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR