(S)-1-Methyl-3-pyrrolidinol 98%, ee 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(S)-3-Hydroxy-1-methylpyrrolidine, (S)-(+)-1-Methyl-3-pyrrolidinol
L19695.MD
L19695.MDEA
65.7
EUR
L19695.MD
(S)-1-Methyl-3-pyrrolidinol 98%, ee 99%
(S)-1-Methyl-3-pyrrolidinol
Purity: 98%
CAS No.: 104641-59-0
Molecular Formula: C5H11NO
Formula Weight: 101.15
Flash Point: 70°(158°F)
Refractive Index: 1.4640
Boiling Point: 103-106°/35mm
Beilstein Reference: 4349394
MDL No.: MFCD03788748
Formel:
C₅H₁₁NO Molecular Weight: 101,15 g/mol Siedepunkt: 103…106 °C (35 mmHg) Dichte: 0,920 Flammpunkt: 70 °C (158 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788748 CAS-Nummer: 104641-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...