(S)-(-)-1-Boc-3-aminopyrrolidin 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98%
CAS No.: 147081-44-5
Molecular Formula: C9H18N2O2
Formula Weight: 186.25
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4715
Boiling Point: 115°/4mm
Beilstein Reference: 8312125
Optical Rotation: -3° (neat)
UN No.: 2735
MDL No.: MFCD03419271
Formel:
C₉H₁₈N₂O₂ Siedepunkt: 115 °C (4 mmHg) Dichte: 1,020 |
MDL:
MFCD03419271 CAS-Nummer: 147081-44-5 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...