trans-1-Octenylboronic acid pinacol ester 97%
Lieferant: Thermo Fisher Scientific
L19697.03
L19697.03EA
65.4
EUR
L19697.03
trans-1-Octenylboronic acid pinacol ester 97%
trans-1-Octenylboronic acid pinacol ester
Purity: 97%
CAS No.: 83947-55-1
Molecular Formula: C14H27BO2
Formula Weight: 238.18
Refractive Index: 1.4460
Beilstein Reference: 7291389
MDL No.: MFCD03453667
Formel:
C₁₄H₂₇BO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03453667 CAS-Nummer: 83947-55-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...