4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Nitro-3-(trifluoromethyl)benzene-1-sulphonyl chloride
L19721.03
L19721.06
L19721.03EA
41.6
EUR
L19721.03
L19721.06
4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid ≥97%
4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid
Purity: 97%
CAS No.: 39234-83-8
Molecular Formula: C7H3ClF3NO4S
Formula Weight: 289.62
Melting Point: 63-65°
Beilstein Reference: 2880842
UN No.: 3261
MDL No.: MFCD03094387
Formel:
C₇H₃ClF₃NO₄S Schmelzpunkt: 63…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094387 CAS-Nummer: 39234-83-8 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...