3,5-Dichlor-2-pyridincarbonitril 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3,5-Dichloropicolinonitrile, 3,5-Dichloro-2-cyanopyridine
L19723.14
L19723.14EA
120
EUR
L19723.14
3,5-Dichlor-2-pyridincarbonitril 95%
3,5-Dichlor-2-pyridincarbonitril
Purity: 95%
CAS No.: 85331-33-5
Molecular Formula: C6H2Cl2N2
Formula Weight: 173.00
Melting Point: 99-103°
Beilstein Reference: 4390101
UN No.: 3439
MDL No.: MFCD03788758
Formel:
C₆H₂Cl₂N₂ Molecular Weight: 173,00 g/mol Schmelzpunkt: 99…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788758 CAS-Nummer: 85331-33-5 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...