2-Amino-3,5-difluorpyridin 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3,5-Difluoro-2-pyridylamine, 3,5-Difluoropyridin-2-amine

L19727.06
L19727.06EA 84 EUR
L19727.06
2-Amino-3,5-difluorpyridin 98%
2-Amino-3,5-difluorpyridin

Purity: 98% CAS No.: 732306-31-9 Molecular Formula: C5H4F2N2 Formula Weight: 130.09 Melting Point: 55-59° UN No.: 2811 MDL No.: MFCD03788760

Formel: C₅H₄F₂N₂
Schmelzpunkt: 55…59 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788760
CAS-Nummer: 732306-31-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR