Magnesium tert-butoxide ≥90%
Lieferant: Thermo Scientific
|
Gefahr
|
L19745.18
L19745.18EA
105
EUR
L19745.18
Magnesium tert-butoxide ≥90%
Magnesium tert-butoxide
Purity: 90+%
CAS No.: 32149-57-8
Molecular Formula: C8H18MgO
Formula Weight: 170.54
Beilstein Reference: 3979348
UN No.: UN2921
MDL No.: MFCD03788761
Formel:
C₈H₁₈MgO₂ Molecular Weight: 170,535 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788761 CAS-Nummer: 32149-57-8 EINECS: 250-931-3 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...