3-Cyan-4-methylpyridin ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Methyl-3-pyridincarbonitril, 3-Cyan-4-picolin
L19769.03
L19769.06
L19769.14
L19769.03EA
37.7
EUR
L19769.03
L19769.06
L19769.14
3-Cyan-4-methylpyridin ≥97%
3-Cyano-4-methylpyridine
Purity: 97%
CAS No.: 5444-01-9
Molecular Formula: C7H6N2
Formula Weight: 118.14
Odour: Characteristic
Melting Point: 41-44°
Boiling Point: 108-111°/20mm
Beilstein Reference: 112016
MDL No.: MFCD00234272
Formel:
C₇H₆N₂ Molecular Weight: 118.14 g/mol Siedepunkt: 108…111 °C (20 mmHg) Schmelzpunkt: 41…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234272 CAS-Nummer: 5444-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...