3-(Trifluormethoxy)phenylboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L19775.03
L19775.06
L19775.03EA
42.2
EUR
L19775.03
L19775.06
3-(Trifluormethoxy)phenylboronsäure ≥98%
3-(Trifluormethoxy)phenylboronsäure
Purity: 98%
CAS No.: 179113-90-7
Molecular Formula: C7H6BF3O3
Formula Weight: 205.93
Melting Point: ca 80°
Beilstein Reference: 7531456
Formel:
C₇H₆BF₃O₃ Schmelzpunkt: ∼ 80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01320697 CAS-Nummer: 179113-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...