3,3-Difluorpyrrolidinhydrochlorid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,3-Difluoropyrrolidine HCl

L19780.ME
L19780.MEEA 934 EUR
L19780.ME
3,3-Difluorpyrrolidinhydrochlorid 98%
3,3-Difluorpyrrolidinhydrochlorid

Purity: 98% CAS No.: 163457-23-6 Molecular Formula: C4H7F2N.HCl Formula Weight: 143.56 Melting Point: 130-134° MDL No.: MFCD03788948

Formel: C₄H₇F₂N
Molecular Weight: 143,56 g/mol
Schmelzpunkt: 130…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788948
CAS-Nummer: 163457-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR