3-(Boc-amino)-3-phenylpropionsäure 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
Boc-DL-β-Phe-OH, 3-tert-Butoxycarbonylamino-3-phenyl-propionic acid
L19802.03
L19802.03EA
74.5
EUR
L19802.03
3-(Boc-amino)-3-phenylpropionsäure 97%
3-(Boc-amino)-3-phenylpropionsäure
Purity: 97%
CAS No.: 14676-01-8
Molecular Formula: C14H19NO4
Formula Weight: 265.31
Storage Temperature: 2°C to 8°C
Melting Point: 142-144° dec.
Beilstein Reference: 2867023
MDL No.: MFCD02090695
Formel:
C₁₄H₁₉NO₄ Molecular Weight: 265.31 g/mol Schmelzpunkt: 142…144 °C (Zers.) |
MDL:
MFCD02090695 CAS-Nummer: 14676-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...