2-Aminoisonicotinamid 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L19843.03
L19843.06
L19843.14
L19843.03EA
57.8
EUR
L19843.03
L19843.06
L19843.14
2-Aminoisonicotinamid 95%
2-Aminoisonicotinamid
Purity: 95%
CAS No.: 13538-42-6
Molecular Formula: C6H7N3O
Formula Weight: 137.14
Melting Point: 252-258° dec.
Beilstein Reference: 115924
Formel:
C₆H₇N₃O Schmelzpunkt: 252…258 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791262 CAS-Nummer: 13538-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...