3-(Trifluormethoxy)thiophenol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

L19912.03 L19912.06
L19912.03EA 82.8 EUR
L19912.03 L19912.06
3-(Trifluormethoxy)thiophenol ≥98%
3-(Trifluormethoxy)thiophenol

Purity: 98% CAS No.: 220239-66-7 Molecular Formula: C7H5F3OS Formula Weight: 194.18 Refractive Index: 1.4770

Formel: C₇H₅F₃OS
Lagertemperatur: Raumtemperatur
MDL: MFCD01320799
CAS-Nummer: 220239-66-7
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR