7-Brom-1-chlorisochinolin ≥97%

Lieferant: Thermo Scientific
Achtung

L19946.06
L19946.06EA 180 EUR
L19946.06
7-Brom-1-chlorisochinolin ≥97%
7-Bromo-1-chloroisoquinoline

Purity: 97% CAS No.: 215453-51-3 Molecular Formula: C9H5BrClN Formula Weight: 242.50 Melting Point: 122-126° Beilstein Reference: 8404252

Formel: C₉H₅BrClN
Molecular Weight: 242,50 g/mol
Schmelzpunkt: 122…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039320
CAS-Nummer: 215453-51-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR