2,6-Bis(trifluormethyl)phenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

L19952.03 L19952.06 L19952.MD
L19952.03EA 80 EUR
L19952.03 L19952.06 L19952.MD
2,6-Bis(trifluormethyl)phenylboronsäure ≥97%
2,6-Bis(trifluormethyl)phenylboronsäure

Purity: 97% CAS No.: 681812-07-7 Molecular Formula: C8H5BF6O2 Formula Weight: 257.93 Melting Point: ca 150° MDL No.: MFCD04039321

Formel: C₈H₅BF₆O₂
Schmelzpunkt: ∼ 150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039321
CAS-Nummer: 681812-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR